Assay Details
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632026.1 | XP_023487794.1 | 728 | 87 | ||
GenBank mRNA | JL618870.1 | - | 2 - 3 | 185 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
