Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500763.5 | XP_001500813.3 | 546 | 80 | ||
XM_005610008.3 | XP_005610065.2 | 563 | 80 | |||
XM_023640954.1 | XP_023496722.1 | 453 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023640954.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
