Assay Details
Gene Symbol: SYT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499240.5 | XP_001499290.1 | 1131 | 80 | ||
XM_023641037.1 | XP_023496805.1 | 1135 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
