Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641084.1 | XP_023496852.1 | 3468 | 83 | ||
XM_023641085.1 | XP_023496853.1 | 3467 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
