Assay Details
Gene Symbol: NRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502149.5 | XP_001502199.4 | 190 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502149.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
