Assay Details
Gene Symbol: KLHL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645709.1 | XP_023501477.1 | 4447 | 93 | ||
XM_023645710.1 | XP_023501478.1 | 1568 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
