Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638342.1 | XP_023494110.1 | 861 | 101 | ||
XM_023638343.1 | XP_023494111.1 | 901 | 101 | |||
XM_023638346.1 | XP_023494114.1 | 863 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
