Assay Details
Gene Symbol: TRUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495127.5 | XP_001495177.1 | 357 | 63 | ||
XM_023638805.1 | XP_023494573.1 | 364 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
