Assay Details
Gene Symbol: CASP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733325.2 | XP_014588811.1 | 805 | 62 | ||
XM_014733326.2 | XP_014588812.1 | 840 | 62 | |||
XM_014733327.2 | XP_014588813.1 | 986 | 62 | |||
XM_014733328.2 | XP_014588814.1 | 1463 | 62 | |||
XM_023639063.1 | XP_023494831.1 | 700 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
