Assay Details
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500281.5 | XP_001500331.1 | 392 | 71 | ||
GenBank mRNA | JL628102.1 | - | 2 - 3 | 267 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
