Assay Details
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500866.4 | XP_001500916.1 | 966 | 70 | ||
XM_005602306.3 | XP_005602363.1 | 1017 | 70 | |||
XM_014733156.2 | XP_014588642.1 | 1017 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
