Assay Details
Gene Symbol: R3HCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501230.5 | XP_001501280.2 | 2225 | 77 | ||
XM_005602317.3 | XP_005602374.1 | 2226 | 77 | |||
XM_014733204.2 | XP_014588690.1 | 2405 | 77 | |||
XM_023641283.1 | XP_023497051.1 | 2199 | 77 | |||
XM_023641287.1 | XP_023497055.1 | 2226 | 77 | |||
XM_023641294.1 | XP_023497062.1 | 608 | 77 | |||
GenBank mRNA | JL620510.1 | - | 2 - 3 | 854 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
