Assay Details
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501483.3 | XP_001501533.2 | 761 | 90 | ||
XM_014733408.2 | XP_014588894.1 | 653 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
