Assay Details
Gene Symbol: MINPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503220.5 | XP_001503270.2 | 677 | 103 | ||
XM_003363467.4 | XP_003363515.2 | 677 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
