Assay Details
Gene Symbol: TBATA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502787.6 | XP_001502837.3 | 1154 | 71 | ||
XM_023644416.1 | XP_023500184.1 | 1157 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
