Assay Details
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503115.5 | XP_001503165.2 | 211 | 80 | ||
XM_003363393.4 | XP_003363441.1 | 211 | 80 | |||
XM_005602512.3 | XP_005602569.1 | 211 | 80 | |||
XM_005602513.3 | XP_005602570.1 | 211 | 80 | |||
XM_005602514.3 | XP_005602571.1 | 211 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
