Assay Details
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647160.1 | XP_023502928.1 | 384 | 142 | ||
XM_023647165.1 | XP_023502933.1 | 490 | 142 | |||
XM_023647171.1 | XP_023502939.1 | 462 | 142 | |||
XM_023647175.1 | XP_023502943.1 | 566 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
