Assay Details
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647429.1 | XP_023503197.1 | 1952 | 85 | ||
XM_023647436.1 | XP_023503204.1 | 2131 | 85 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647429.1 | XP_023503197.1 | 1952 | 85 | ||
XM_023647436.1 | XP_023503204.1 | 2131 | 85 |