Assay Details
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499219.6 | XP_001499269.2 | 157 | 57 | ||
XM_023649898.1 | XP_023505666.1 | 157 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
