Assay Details
Gene Symbol: MRPL46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916648.4 | XP_001916683.1 | 369 | 65 | ||
XM_023650346.1 | XP_023506114.1 | 369 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
