Assay Details
Gene Symbol: THEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739882.2 | XP_014595368.1 | 1863 | 63 | ||
XM_023641155.1 | XP_023496923.1 | 1863 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
