Assay Details
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497050.2 | XP_001497100.2 | 367 | 77 | ||
XM_014740197.2 | XP_014595683.1 | 345 | 77 | |||
XM_014740199.2 | XP_014595685.1 | 368 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
