Assay Details
Gene Symbol: CD101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496566.5 | XP_001496616.2 | 4686 | 71 | ||
XM_014740202.2 | XP_014595688.1 | 4944 | 71 | |||
XM_023641343.1 | XP_023497111.1 | 5889 | 71 | |||
XM_023641344.1 | XP_023497112.1 | 3714 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
