Assay Details
Gene Symbol: GABRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652053.1 | XP_023507821.1 | 1127 | 69 | ||
XM_023652057.1 | XP_023507825.1 | 1304 | 69 | |||
XM_023652061.1 | XP_023507829.1 | 1290 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
