Assay Details
Gene Symbol: PPCDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491716.5 | XP_001491766.2 | 886 | 87 | ||
XM_005602910.3 | XP_005602967.1 | 1737 | 87 | |||
XM_005602911.3 | XP_005602968.1 | 1572 | 87 | |||
XM_014733931.2 | XP_014589417.1 | 677 | 87 | |||
XM_023653397.1 | XP_023509165.1 | 701 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014733931.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
