Assay Details
Gene Symbol: LOXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917702.5 | XP_001917737.3 | 1970 | 59 | ||
XM_023653958.1 | XP_023509726.1 | 814 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
