Assay Details
Gene Symbol: CD276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654011.1 | XP_023509779.1 | 383 | 73 | ||
XM_023654016.1 | XP_023509784.1 | 651 | 73 | |||
XM_023654020.1 | XP_023509788.1 | 414 | 73 | |||
XM_023654027.1 | XP_023509795.1 | 270 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
