Assay Details
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654752.1 | XP_023510520.1 | 286 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023654752.1 transcripts.
Set Membership: |
![]() ![]() |
