Assay Details
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500219.4 | XP_001500269.1 | 591 | 79 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500219.4 | XP_001500269.1 | 591 | 79 |