Assay Details
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503635.5 | XP_001503685.1 | 603 | 66 | ||
XM_014734065.2 | XP_014589551.1 | 610 | 66 | |||
XM_023620455.1 | XP_023476223.1 | 202 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
