Assay Details
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503656.6 | XP_001503706.4 | 1043 | 83 | ||
XM_023620582.1 | XP_023476350.1 | 1043 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
