Assay Details
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622641.1 | XP_023478409.1 | 1179 | 62 | ||
XM_023622646.1 | XP_023478414.1 | 1179 | 62 | |||
XM_023622654.1 | XP_023478422.1 | 1179 | 62 | |||
XM_023622663.1 | XP_023478431.1 | 1179 | 62 | |||
XM_023622672.1 | XP_023478440.1 | 1179 | 62 | |||
XM_023622678.1 | XP_023478446.1 | 1179 | 62 | |||
XM_023622686.1 | XP_023478454.1 | 1179 | 62 | |||
XM_023622697.1 | XP_023478465.1 | 1179 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
