Assay Details
Gene Symbol: TSSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363558.4 | XP_003363606.1 | 581 | 93 | ||
XM_014733724.2 | XP_014589210.1 | 581 | 93 | |||
XM_023622928.1 | XP_023478696.1 | 940 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
