Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489087.4 | XP_001489137.2 | 2078 | 69 | ||
XM_023623063.1 | XP_023478831.1 | 2080 | 69 | |||
XM_023623070.1 | XP_023478838.1 | 1037 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
