Assay Details
Gene Symbol: NPAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623732.1 | XP_023479500.1 | 707 | 106 | ||
XM_023623740.1 | XP_023479508.1 | 701 | 106 | |||
XM_023623744.1 | XP_023479512.1 | 707 | 106 | |||
XM_023623775.1 | XP_023479543.1 | 656 | 106 | |||
XM_023623780.1 | XP_023479548.1 | 650 | 106 | |||
XM_023623784.1 | XP_023479552.1 | 566 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
