Assay Details
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603434.3 | XP_005603491.1 | 779 | 94 | ||
XM_023623889.1 | XP_023479657.1 | 461 | 94 | |||
GenBank mRNA | JL616451.2 | - | 1 - 2 | 264 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
