Assay Details
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491270.4 | XP_001491320.1 | 342 | 113 | ||
XM_023624004.1 | XP_023479772.1 | 260 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
