Assay Details
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624599.1 | XP_023480367.1 | 256 | 87 | ||
XM_023624604.1 | XP_023480372.1 | 257 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
