Assay Details
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649507.1 | XP_023505275.1 | 452 | 66 | ||
XM_023649508.1 | XP_023505276.1 | 452 | 66 | |||
XM_023649509.1 | XP_023505277.1 | 452 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
