Assay Details
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362228.4 | XP_003362276.1 | 433 | 54 | ||
XM_005596145.1 | XP_005596202.1 | 419 | 54 | |||
XM_005596146.3 | XP_005596203.1 | 413 | 54 | |||
XM_023649537.1 | XP_023505305.1 | 512 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
