Assay Details
Gene Symbol: THAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649610.1 | XP_023505378.1 | 135 | 94 | ||
XM_023649611.1 | XP_023505379.1 | 135 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023649610.1, XM_023649611.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
