Assay Details
Gene Symbol: SPINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496775.6 | XP_001496825.3 | 998 | 57 | ||
XM_023649633.1 | XP_023505401.1 | 938 | 57 | |||
XM_023649634.1 | XP_023505402.1 | 884 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
