Assay Details
Gene Symbol: PAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649695.1 | XP_023505463.1 | 485 | 67 | ||
XM_023649697.1 | XP_023505465.1 | 697 | 67 | |||
XM_023649698.1 | XP_023505466.1 | 358 | 67 | |||
XM_023649699.1 | XP_023505467.1 | 498 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
