Assay Details
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501566.5 | XP_001501616.1 | 149 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501566.5 transcripts.
Set Membership: |
![]() ![]() |
