Assay Details
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649936.1 | XP_023505704.1 | 2068 | 89 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649936.1 | XP_023505704.1 | 2068 | 89 |