Assay Details
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650140.1 | XP_023505908.1 | 1020 | 77 | ||
XM_023650141.1 | XP_023505909.1 | 1018 | 77 | |||
XM_023650142.1 | XP_023505910.1 | 1018 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
