Assay Details
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917755.5 | XP_001917790.5 | 848 | 71 | ||
XM_014729833.2 | XP_014585319.2 | 849 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
