Assay Details
Gene Symbol: PTOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596409.3 | XP_005596466.1 | 1488 | 61 | ||
XM_023650189.1 | XP_023505957.1 | 1488 | 61 | |||
XM_023650190.1 | XP_023505958.1 | 1053 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
