Assay Details
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650226.1 | XP_023505994.1 | 211 | 115 | ||
XM_023650227.1 | XP_023505995.1 | 212 | 115 | |||
XM_023650228.1 | XP_023505996.1 | 64 | 115 |

More Information
Set Membership: |
![]() ![]() |
