Assay Details
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650226.1 | XP_023505994.1 | 578 | 88 | ||
XM_023650227.1 | XP_023505995.1 | 558 | 88 | |||
XM_023650228.1 | XP_023505996.1 | 396 | 88 | |||
XM_023650229.1 | XP_023505997.1 | 216 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
